Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r13b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011625.1 | NP_035755.1 | 11 - 12 | 2000 | 84 | |
GenBank mRNA | BC034207.1 | - | 7 - 8 | 1446 | 84 | |
BC053092.1 | - | 11 - 12 | 2026 | 5719383 | 84 | |
BC053732.1 | - | 7 - 8 | 1446 | 84 | ||
BC054788.1 | - | 11 - 12 | 2000 | 6826065 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |