Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mctp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030174.2 | NP_084450.2 | 18 - 19 | 2156 | 71 | |
GenBank mRNA | AK047562.1 | - | 18 - 19 | 2157 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |