Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bbx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027444.3 | NP_081720.2 | 2 - 3 | 222 | 61 | |
GenBank mRNA | AF276950.1 | - | 2 - 3 | 107 | 61 | |
AF276951.1 | - | 2 - 3 | 194 | 61 | ||
AF454943.1 | - | 2 - 3 | 215 | 61 | ||
AK017487.1 | - | 2 - 3 | 215 | 61 | ||
AK029747.1 | - | 2 - 3 | 222 | 61 | ||
AK049516.1 | - | 2 - 3 | 208 | 61 | ||
AK050488.1 | - | 2 - 3 | 161 | 61 | ||
AK157813.1 | - | 2 - 3 | 201 | 61 | ||
AK167724.1 | - | 2 - 3 | 353 | 61 | ||
BC019456.1 | - | 2 - 3 | 297 | 61 | ||
BC057869.1 | - | 2 - 3 | 190 | 61 | ||
BC066821.1 | - | 2 - 3 | 173 | 30061364 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |