Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400020989.mm.1 |
TC0400002572.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400020989.mm.1 |
NM_001099675.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020989.mm.1 |
NM_001099675.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020989.mm.1 |
NM_001134646.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020989.mm.1 |
NM_001134646.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020984.mm.1 |
TC0400002572.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400020984.mm.1 |
NM_001099675.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020984.mm.1 |
NM_001099675.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020984.mm.1 |
NM_001134646.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020984.mm.1 |
NM_001134646.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020988.mm.1 |
TC0400002572.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400020988.mm.1 |
NM_001099675.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020988.mm.1 |
NM_001099675.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020988.mm.1 |
NM_001134646.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020988.mm.1 |
NM_001134646.1 |
Refseq |