Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csgalnact1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252623.1 | NP_001239552.1 | 5 - 6 | 1636 | 95 | |
NM_172753.5 | NP_766341.4 | 4 - 5 | 1332 | 95 | ||
GenBank mRNA | AK028688.1 | - | 4 - 5 | 1255 | 95 | |
AK033522.1 | - | 2 - 3 | 986 | 95 | ||
AK049770.1 | - | 5 - 6 | 1930 | 95 | ||
AK080687.1 | - | 4 - 5 | 1301 | 95 | ||
AK133567.1 | - | 2 - 3 | 985 | 95 | ||
AK144089.1 | - | 1 - 2 | 893 | 95 | ||
BC057630.1 | - | 6 - 7 | 1604 | 95 | ||
BC089162.1 | - | 4 - 5 | 1329 | 30470205 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |