Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shroom3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077595.2 | NP_001071063.1 | 6 - 7 | 4514 | 75 | |
NM_001077596.2 | NP_001071064.1 | 6 - 7 | 4562 | 75 | ||
NM_015756.2 | NP_056571.2 | 7 - 8 | 4886 | 75 | ||
GenBank mRNA | AF199421.1 | - | 7 - 8 | 4904 | 75 | |
AF199422.1 | - | 6 - 7 | 4472 | 75 | ||
AK049347.1 | - | 3 - 4 | 2420 | 75 | ||
AK129371.1 | - | 3 - 4 | 3892 | 75 | ||
AK147493.1 | - | 6 - 7 | 4497 | 75 | ||
AK164918.1 | - | 6 - 7 | 4473 | 75 | ||
BC003909.1 | - | 1 - 2 | 158 | 75 | ||
BC145113.1 | - | 8 - 9 | 4303 | 9053661 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |