Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btnl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138678.2 | NP_619619.2 | 2 - 3 | 220 | 66 | |
GenBank mRNA | AF269232.1 | - | 2 - 3 | 220 | 66 | |
AK029784.1 | - | 2 - 3 | 326 | 66 | ||
AK143638.1 | - | 2 - 3 | 370 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |