Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rufy3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289774.1 | NP_001276703.1 | 14 - 15 | 1812 | 137 | |
GenBank mRNA | AK154941.1 | - | 14 - 15 | 1813 | 137 | |
BP765560.1 | - | 4 - 5 | 417 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |