Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gatad2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113346.1 | NP_001106817.1 | 4 - 5 | 793 | 69 | |
NM_001286450.1 | NP_001273379.1 | 4 - 5 | 941 | 69 | ||
NM_145596.4 | NP_663571.2 | 4 - 5 | 941 | 69 | ||
GenBank mRNA | AK089074.1 | - | 4 - 5 | 923 | 69 | |
AK160058.1 | - | 4 - 5 | 790 | 69 | ||
BC038221.1 | - | 4 - 5 | 691 | 5321913 | 69 | |
CN666280.1 | - | 2 - 3 | 311 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |