Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283028.1 | NP_001269957.1 | 3 - 4 | 441 | 84 | |
NM_001283029.1 | NP_001269958.1 | 3 - 4 | 306 | 84 | ||
NM_001283030.1 | NP_001269959.1 | 2 - 3 | 165 | 84 | ||
NM_146043.4 | NP_666155.1 | 3 - 4 | 366 | 84 | ||
GenBank mRNA | AK088127.1 | - | 3 - 4 | 307 | 84 | |
AK134259.1 | - | 3 - 4 | 329 | 84 | ||
AK168544.1 | - | 3 - 4 | 367 | 84 | ||
BC016517.1 | - | 3 - 4 | 352 | 2654233 | 84 | |
CJ173506.1 | - | 3 - 4 | 425 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |