Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175401.3 | NP_780610.2 | 2 - 3 | 145 | 59 | |
GenBank mRNA | AK135553.1 | - | 2 - 3 | 159 | 59 | |
BC040428.1 | - | 2 - 3 | 145 | 3327345 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |