Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175401.3 | NP_780610.2 | 5 - 6 | 640 | 82 | |
GenBank mRNA | AK135553.1 | - | 5 - 6 | 654 | 82 | |
AK155157.1 | - | 5 - 6 | 739 | 82 | ||
BC040428.1 | - | 5 - 6 | 640 | 3327345 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |