Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134071.3 | NP_598832.3 | 6 - 7 | 841 | 168 | |
GenBank mRNA | AK089501.1 | - | 5 - 6 | 678 | 169 | |
AK171473.1 | - | 5 - 6 | 678 | 168 | ||
BC061188.1 | - | 6 - 7 | 821 | 168 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |