Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028384.1 | NP_082660.1 | 10 - 11 | 1427 | 97 | |
GenBank mRNA | BM949737.1 | - | 3 - 4 | 515 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |