Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023281.1 | NP_075770.1 | 13 - 14 | 1830 | 63 | |
GenBank mRNA | AK004362.1 | - | 3 - 4 | 381 | 63 | |
AK010561.1 | - | 13 - 14 | 1820 | 63 | ||
AK029520.1 | - | 13 - 14 | 1830 | 63 | ||
AK034928.1 | - | 13 - 14 | 1831 | 63 | ||
AK049590.1 | - | 13 - 14 | 1817 | 63 | ||
AK050475.1 | - | 13 - 14 | 1817 | 63 | ||
AK075990.1 | - | 13 - 14 | 1814 | 63 | ||
AK145923.1 | - | 13 - 14 | 1828 | 63 | ||
AK147286.1 | - | 13 - 14 | 1817 | 63 | ||
AK147624.1 | - | 13 - 14 | 1817 | 63 | ||
AK151333.1 | - | 5 - 6 | 458 | 63 | ||
AK153085.1 | - | 13 - 14 | 1837 | 63 | ||
AK162148.1 | - | 13 - 14 | 1813 | 63 | ||
AK169254.1 | - | 13 - 14 | 1813 | 63 | ||
BC011301.1 | - | 13 - 14 | 1790 | 63 | ||
BC031849.1 | - | 13 - 14 | 1838 | 5067510 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |