Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahrr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009644.2 | NP_033774.1 | 5 - 6 | 517 | 98 | |
GenBank mRNA | AB015140.1 | - | 5 - 6 | 517 | 98 | |
AK154152.1 | - | 5 - 6 | 506 | 98 | ||
AK154761.1 | - | 5 - 6 | 539 | 98 | ||
AK155711.1 | - | 5 - 6 | 539 | 98 | ||
AK173134.1 | - | 2 - 3 | 159 | 98 | ||
BC118534.1 | - | 4 - 5 | 323 | 40095111 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |