Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028947.1 | NP_083223.1 | 6 - 7 | 845 | 67 | |
GenBank mRNA | AK016872.1 | - | 6 - 7 | 846 | 67 | |
BC100522.1 | - | 6 - 7 | 743 | 30920038 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |