Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400031609.mm.1 |
TC0400003832.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400031609.mm.1 |
NM_011110.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400031609.mm.1 |
NM_011110.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400031612.mm.1 |
TC0400003832.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400031612.mm.1 |
NM_001122954.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400031612.mm.1 |
NM_001122954.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400031606.mm.1 |
TC0400003832.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400031606.mm.1 |
NM_001122954.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400031606.mm.1 |
NM_001122954.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400031606.mm.1 |
NM_011110.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400031606.mm.1 |
NM_011110.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400031601.mm.1 |
TC0400003832.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400031601.mm.1 |
NM_001122954.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400031601.mm.1 |
NM_001122954.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400031601.mm.1 |
NM_011110.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400031601.mm.1 |
NM_011110.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400031607.mm.1 |
TC0400003832.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400031607.mm.1 |
NM_011110.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400031607.mm.1 |
NM_011110.4 |
Refseq |