Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp3a57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100180.1 | NP_001093650.1 | 2 - 3 | 242 | 157 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |