Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skint10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177668.2 | NP_808336.1 | 1 - 2 | 106 | 119 | |
GenBank mRNA | AK037152.1 | - | 1 - 2 | 71 | 119 | |
EF494905.1 | - | 1 - 2 | 69 | 119 | ||
EF494906.1 | - | 1 - 2 | 69 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |