Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026533.3 | NP_080809.1 | 2 - 3 | 162 | 154 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Inventoried Probe spans exons |