Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hephl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164797.1 | NP_001158269.1 | 12 - 13 | 2297 | 77 | |
GenBank mRNA | AK132457.1 | - | 12 - 13 | 2298 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |