Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008972.2 | NP_032998.1 | 1 - 2 | 217 | 61 | |
GenBank mRNA | AK037980.1 | - | 1 - 2 | 228 | 61 | |
AK084218.1 | - | 1 - 2 | 227 | 61 | ||
AK088005.1 | - | 1 - 2 | 228 | 61 | ||
AK132003.1 | - | 1 - 2 | 234 | 61 | ||
AK142076.1 | - | 1 - 2 | 233 | 61 | ||
AK143576.1 | - | 1 - 2 | 228 | 61 | ||
AK145650.1 | - | 1 - 2 | 228 | 61 | ||
AK146454.1 | - | 1 - 2 | 228 | 61 | ||
AK146651.1 | - | 1 - 2 | 228 | 61 | ||
AK146849.1 | - | 1 - 2 | 228 | 61 | ||
AK151289.1 | - | 1 - 2 | 227 | 61 | ||
AK151511.1 | - | 1 - 2 | 231 | 61 | ||
AK151768.1 | - | 1 - 2 | 228 | 61 | ||
AK152032.1 | - | 1 - 2 | 227 | 61 | ||
AK153178.1 | - | 1 - 2 | 228 | 61 | ||
AK153496.1 | - | 1 - 2 | 228 | 61 | ||
AK153953.1 | - | 1 - 2 | 232 | 61 | ||
AK159128.1 | - | 1 - 2 | 228 | 61 | ||
BC081453.1 | - | 1 - 2 | 217 | 6820943 | 61 | |
BC083135.1 | - | 1 - 2 | 180 | 6506450 | 61 | |
BC085171.1 | - | 1 - 2 | 217 | 30699339 | 61 | |
X56135.1 | - | 1 - 2 | 217 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |