Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033408.4 | NP_001028580.1 | 4 - 5 | 1715 | 111 | |
GenBank mRNA | AK133355.1 | - | 4 - 5 | 1716 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |