Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pate2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033421.3 | NP_001028593.1 | 2 - 3 | 121 | 128 | |
GenBank mRNA | AK136651.1 | - | 2 - 3 | 122 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |