Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037927.1 | NP_001033016.1 | 15 - 16 | 1702 | 102 | |
GenBank mRNA | AB205021.1 | - | 15 - 16 | 1702 | 102 | |
BC150920.1 | - | 14 - 15 | 1672 | 9087831 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |