Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tuft1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293728.1 | NP_001280657.1 | 9 - 10 | 941 | 75 | |
NM_011656.3 | NP_035786.1 | 10 - 11 | 1016 | 75 | ||
GenBank mRNA | AF002860.1 | - | 10 - 11 | 910 | 75 | |
AF047704.1 | - | 10 - 11 | 950 | 75 | ||
AK132488.1 | - | 10 - 11 | 948 | 75 | ||
AK171705.1 | - | 9 - 10 | 906 | 75 | ||
BC019213.1 | - | 10 - 11 | 947 | 5064658 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |