Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dytn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081658.1 | NP_001075127.1 | 5 - 6 | 683 | 90 | |
GenBank mRNA | DQ443727.1 | - | 5 - 6 | 683 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |