Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pttg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001131054.1 | NP_001124526.1 | 3 - 4 | 330 | 109 | |
NM_013917.2 | NP_038945.2 | 2 - 3 | 582 | 109 | ||
GenBank mRNA | AF071209.1 | - | 2 - 3 | 582 | 109 | |
AK002473.1 | - | 2 - 3 | 296 | 109 | ||
AK008704.1 | - | 2 - 3 | 297 | 109 | ||
AK088387.1 | - | 2 - 3 | 296 | 109 | ||
BC023324.1 | - | 3 - 4 | 336 | 4946234 | 109 | |
CN837307.1 | - | 3 - 4 | 335 | 109 | ||
DV657646.1 | - | 2 - 3 | 300 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |