Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arglu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK034581.1 | - | 1944 | 179 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ephrin B2 | Efnb2 |
ELF-2, Epl5, Eplg5, Htk-L, LERK-5, Lerk5, NLERK-1 |
-4.3kb | Sense | Chr. 8 - 8617235 - 8660773 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |