Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlgn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK158508.1 | - | 1 | 348 | 72 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | N-acetylated alpha-linked acidic dipeptidase-like 2 | Naaladl2 |
2810043G22Rik, EG635702, Gm1021 |
-1.4mb | Sense | Chr. 3 - 23798103 - 24784006 on NCBI Build GRCm38 |
Downstream | RIKEN cDNA A830092H15 gene | A830092H15Rik | 178.0kb | Anti-sense | Chr. 3 - 26331953 - 26336870 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |