Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100029572.mm.1 |
NM_001098203.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100029572.mm.1 |
NM_001098203.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100029572.mm.1 |
NM_010430.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100029572.mm.1 |
NM_010430.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100029572.mm.1 |
TC1100003217.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100029571.mm.1 |
NM_001098203.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100029571.mm.1 |
NM_001098203.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100029571.mm.1 |
NM_010430.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100029571.mm.1 |
NM_010430.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100029571.mm.1 |
TC1100003217.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100029573.mm.1 |
NM_001098203.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100029573.mm.1 |
NM_001098203.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100029573.mm.1 |
TC1100003217.mm.1 |
MTA TC |