Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103171.1 | NP_001096641.1 | 59 - 60 | 9968 | 85 | |
NM_010862.2 | NP_034992.2 | 60 - 61 | 10022 | 85 | ||
NM_182698.2 | NP_874357.2 | 58 - 59 | 6206 | 85 | ||
GenBank mRNA | AF144095.1 | - | 60 - 61 | 10022 | 85 | |
AK030528.1 | - | 11 - 12 | 1053 | 85 | ||
AY331132.1 | - | 59 - 60 | 9968 | 85 | ||
AY331133.1 | - | 58 - 59 | 6206 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |