Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfaip8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134131.2 | NP_598892.2 | 1 - 2 | 342 | 76 | |
GenBank mRNA | AK087478.1 | - | 1 - 2 | 260 | 76 | |
AK090316.1 | - | 1 - 2 | 152 | 76 | ||
AK090350.1 | - | 1 - 2 | 228 | 76 | ||
AK149901.1 | - | 1 - 2 | 118 | 76 | ||
AK150445.1 | - | 1 - 2 | 114 | 76 | ||
AK151622.1 | - | 1 - 2 | 114 | 76 | ||
AK152426.1 | - | 1 - 2 | 114 | 76 | ||
BC009090.1 | - | 1 - 2 | 104 | 3965693 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |