Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pbx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008783.3 | NP_032809.1 | 7 - 8 | 1749 | 97 | |
GenBank mRNA | AK087587.1 | - | 6 - 7 | 873 | 97 | |
BC054376.1 | - | 7 - 8 | 1710 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons |