Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgals2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025622.3 | NP_079898.2 | 2 - 3 | 137 | 95 | |
GenBank mRNA | AK007364.1 | - | 2 - 3 | 134 | 95 | |
AK008423.1 | - | 2 - 3 | 138 | 95 | ||
AK008607.1 | - | 2 - 3 | 139 | 95 | ||
AK008636.1 | - | 2 - 3 | 138 | 95 | ||
AK008648.1 | - | 2 - 3 | 138 | 95 | ||
BC027504.1 | - | 2 - 3 | 104 | 1434603 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |