Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lamp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177356.3 | NP_796330.2 | 1 - 2 | 149 | 97 | |
GenBank mRNA | AJ510130.1 | - | 1 - 2 | 68 | 97 | |
AK028039.1 | - | 1 - 2 | 101 | 97 | ||
AK033379.1 | - | 1 - 2 | 101 | 97 | ||
BY067461.1 | - | 1 - 2 | 159 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |