Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252519.1 | NP_001239448.1 | 4 - 5 | 403 | 85 | |
NM_001253385.1 | NP_001240314.1 | 4 - 5 | 403 | 85 | ||
NM_177889.5 | NP_808557.2 | 4 - 5 | 403 | 85 | ||
GenBank mRNA | AK037211.1 | - | 4 - 5 | 400 | 85 | |
BC060530.1 | - | 3 - 4 | 330 | 5684819 | 85 | |
CD806198.1 | - | 3 - 4 | 354 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |