Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700002726.mm.1 |
TC0700000376.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700002726.mm.1 |
NM_009567.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002726.mm.1 |
NM_009567.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002722.mm.1 |
TC0700000376.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700002722.mm.1 |
NM_009567.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002722.mm.1 |
NM_009567.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002723.mm.1 |
TC0700000376.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700002723.mm.1 |
NM_009567.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002723.mm.1 |
NM_009567.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002729.mm.1 |
TC0700000376.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700002729.mm.1 |
NM_009567.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002729.mm.1 |
NM_009567.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002728.mm.1 |
TC0700000376.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700002728.mm.1 |
NM_009567.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002728.mm.1 |
NM_009567.4 |
Refseq |