Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dffa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_104263.1 | 2 - 3 | 323 | 84 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | peroxisomal biogenesis factor 14 | Pex14 |
Pex14p, R75137 |
-6.3kb | Anti-sense | Chr. 4 - 148960535 - 149099812 on NCBI Build GRCm38 |
Downstream | cortistatin | Cort |
CST, PCST |
17.3kb | Anti-sense | Chr. 4 - 149125191 - 149126741 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons |