Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SS18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012103457.3 | XP_011958847.2 | 7 - 8 | 891 | 75 | |
XM_015103616.2 | XP_014959102.2 | 7 - 8 | 927 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |