Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134782.1 | NP_001128254.1 | 2 - 3 | 656 | 84 | |
GenBank mRNA | EU925138.1 | - | 2 - 3 | 656 | 84 |
Gene Symbol: LOC114113964 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027967172.1 | XP_027822973.1 | 2 - 3 | 714 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Ovis aries |