Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL6IP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004691.4 | XP_004004740.1 | 1 - 2 | 465 | 92 | |
XM_027964953.1 | XP_027820754.1 | 1 - 2 | 1119 | 92 | ||
XM_027964954.1 | XP_027820755.1 | 1 - 2 | 471 | 92 | ||
XM_027964955.1 | XP_027820756.1 | 1 - 2 | 471 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Ovis aries |