Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC90B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004019415.4 | XP_004019464.2 | 6 - 7 | 948 | 86 | |
XM_027960121.1 | XP_027815922.1 | 5 - 6 | 681 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Ovis aries |