Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4GALT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246211.1 | NP_001233140.1 | 4 - 5 | 994 | 112 | |
GenBank mRNA | JF510320.1 | - | 4 - 5 | 994 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |