Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARFIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162549.1 | NP_001156021.1 | 2 - 3 | 195 | 89 | |
XM_012095242.3 | XP_011950632.1 | 3 - 4 | 327 | 89 | ||
XM_027978975.1 | XP_027834776.1 | 3 - 4 | 295 | 89 | ||
GenBank mRNA | FJ969418.1 | - | 2 - 3 | 195 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |