Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL2RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009415.1 | NP_001009415.1 | 3 - 4 | 605 | 55 | |
GenBank mRNA | X60149.1 | - | 466 | 55 | ||
Z11560.1 | - | 605 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |