Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFAND5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163605.1 | NP_001157077.1 | 1 - 2 | 151 | 98 | |
XM_015092739.2 | XP_014948225.1 | 2 - 3 | 687 | 98 | ||
XM_027963869.1 | XP_027819670.1 | 2 - 3 | 771 | 98 | ||
GenBank mRNA | FJ937959.1 | - | 1 - 2 | 151 | 98 |