Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267883.1 | NP_001254812.1 | 3 - 4 | 350 | 103 | |
GenBank mRNA | JQ320282.1 | - | 3 - 4 | 350 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |