Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGFBP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129733.1 | NP_001123205.1 | 3 - 4 | 699 | 75 | |
XM_012110889.3 | XP_011966279.1 | 3 - 4 | 1666 | 75 | ||
GenBank mRNA | EU649694.1 | - | 4 - 5 | 1460 | 75 | |
EU727460.1 | - | 3 - 4 | 699 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |